Anti-GARS Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: GARS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77855-805
- 77737-982
- Antigen symbol:
- GARS
- GARS
- Antigen synonyms:
- GARS1|Diadenosine tetraphosphate synthetase|EC 2.7.7.-|GlyRS|Glycyl-tRNA synthetase 1|Ap4A synthetase|EC 6.1.1.14|Glycyl-tRNA synthetase|GARSGlycine--tRNA ligase
- GARS1|GARSGlycine--tRNA ligase|EC 6.1.1.14|Diadenosine tetraphosphate synthetase|Ap4A synthetase|EC 2.7.7.-|Glycyl-tRNA synthetase|GlyRS|Glycyl-tRNA synthetase 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P41250
- P41250
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Glycine--tRNA ligase protein (1-389AA)
- Recombinant Human Glycine--tRNA ligase protein (1-389AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together