Anti-PSMA7 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PSMA7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-330
- 77742-591
- Antigen symbol:
- PSMA7
- PSMA7
- Antigen synonyms:
- Proteasome subunit XAPC7|OTTHUMP00000031449|Proteasome subunit RC6-1|PSMA7|XAPC7|OTTHUMP00000031453|RC6 1|OTTHUMP00000031450|Proteasome subunit alpha type 7|PSMA 7|PSA7_HUMAN|Proteasome subunit RC6 1|C6|MGC3755|HSPC|Proteasome (prosome macropain) subunit alpha type 7|Proteasome alpha 7 subunit|Proteasome subunit alpha 4|Proteasome subunit alpha type-7
- C6|HSPC|MGC3755|OTTHUMP00000031449|OTTHUMP00000031450|OTTHUMP00000031453|Proteasome (prosome macropain) subunit alpha type 7|Proteasome alpha 7 subunit|Proteasome subunit alpha 4|Proteasome subunit alpha type 7|Proteasome subunit alpha type-7|Proteasome subunit RC6 1|Proteasome subunit RC6-1|Proteasome subunit XAPC7|PSA7_HUMAN|PSMA 7|PSMA7|RC6 1|XAPC7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14818
- O14818
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Proteasome subunit alpha type-7 protein (5-232AA)
- Recombinant Human Proteasome subunit alpha type-7 protein (5-232AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together