Anti-INTS6 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
0
Type:
Antigen: INTS6
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77741-134
- 77858-147
- Antigen symbol:
- INTS6
- INTS6
- Antigen synonyms:
- DBI 1| DBI-1| DBI1| DDX26| DDX26A| DEAD box protein| DEAD/H (Asp Glu Ala Asp/His) box polypeptide 26| Deleted in cancer 1| DICE1| DKFZp434B105| HDB| Int6| INT6_HUMAN| Integrator complex subunit 6| INTS 6| Notchl2| OTTHUMP00000018439| OTTHUMP00000215034| OTTHUMP00000215036| Protein DDX26| Protein deleted in cancer 1| RNA helicase HDB
- DBI 1|DBI-1|DBI1|DDX26|DDX26A|DEAD box protein|DEAD/H (Asp Glu Ala Asp/His) box polypeptide 26|Deleted in cancer 1|DICE1|DKFZp434B105|HDB|Int6|INT6_HUMAN|Integrator complex subunit 6|INTS 6|Notchl2|OTTHUMP00000018439|OTTHUMP00000215034|OTTHUMP00000215036|Protein DDX26|Protein deleted in cancer 1|RNA helicase HDB
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UL03
- Q9UL03
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Integrator complex subunit 6 protein (571-795AA)
- Recombinant Human Integrator complex subunit 6 protein (571-795AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Spécifications
Frequently Bought Together