Anti-FAM3C Rabbit Polyclonal Antibody
77736-672
New Product
- Antigen symbol:FAM3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q92520
- Antigen synonyms:Fam3c|FAM3C_HUMAN|Family with sequence similarity 3 member C|GS3786|ILEI|Interleukin-like EMT inducer|Predicted osteoblast protein|Protein FAM3C
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Protein FAM3C
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: FAM3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat