Anti-MAPK3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: MAPK3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77742-749
- 77859-465
- Antigen symbol:
- MAPK3
- MAPK3
- Antigen synonyms:
- ERK 1|ERK-1|ERK1|ERT 2|ERT2|Extracellular Signal Regulated Kinase 1|Extracellular signal related kinase 1|Extracellular signal-regulated kinase 1|HGNC6877|HS44KDAP|HUMKER1A|Insulin Stimulated MAP2 Kinase|Insulin-stimulated MAP2 kinase|MAP kinase 1|MAP kinase 3|MAP Kinase|MAP kinase isoform p44|MAPK 1|MAPK 3|MAPK|MAPK1|Mapk3|MGC20180|Microtubule Associated Protein 2 Kinase|Microtubule-associated protein 2 kinase|Mitogen Activated Protein Kinase 3|Mitogen-activated protein kinase 1|Mitogen-activated protein kinase 3|MK03_HUMAN|OTTHUMP00000174538|OTTHUMP00000174541|p44 ERK1|p44 MAPK|p44-ERK1|p44-MAPK|P44ERK1|P44MAPK|PRKM 3|PRKM3|Protein Kinase Mitogen Activated 3
- Insulin-stimulated MAP2 kinase|Mapk3|p44-MAPK|MK03_HUMAN|PRKM3|ERT2|MAPK|Microtubule Associated Protein 2 Kinase|Mitogen Activated Protein Kinase 3|Extracellular Signal Regulated Kinase 1|MGC20180|ERK-1|HUMKER1A|MAPK1|MAP kinase 3|HGNC6877|Insulin Stimulated MAP2 Kinase|MAP kinase 1|Mitogen-activated protein kinase 1|OTTHUMP00000174541|Mitogen-activated protein kinase 3|MAP kinase isoform p44|MAPK 3|MAPK 1|PRKM 3|p44 MAPK|Microtubule-associated protein 2 kinase|P44MAPK|Protein Kinase Mitogen Activated 3|Extracellular signal-regulated kinase 1|MAP Kinase|ERK1|HS44KDAP|OTTHUMP00000174538|P44ERK1|ERT 2|Extracellular signal related kinase 1|p44 ERK1|p44-ERK1|ERK 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P27361
- P27361
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mitogen-activated protein kinase 3 protein (11-379AA)
- Recombinant Human Mitogen-activated protein kinase 3 protein (11-379AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together