Anti-FOXP4 Rabbit Polyclonal Antibody
Catalog # 77737-482
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:FOXP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8IVH2
- Antigen synonyms:1200010K03Rik|2310007G05Rik|FKHLA|FLJ40908|FLJ44184|Fork head related protein like A|Fork head-related protein-like A|Forkhead box P4|Forkhead box protein P4|FOX P4|FOXP 4|foxp4|FOXP4_HUMAN|hFKHLA|mFKHLA|OTTHUMP00000016374|OTTHUMP00000039784|OTTHUMP00000043412|OTTHUMP00000043536|Winged helix repressor FOX P4|Winged helix repressor FOXP 4|Winged helix repressor FOXP4
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Winged helix repressor FOXP4
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: FOXP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat