Anti-HPDL Rabbit Polyclonal Antibody
Catalogue # 77740-347
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:HPDL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96IR7
- Antigen synonyms:4 HPPD L|4-hydroxyphenylpyruvate dioxygenase-like|4-hydroxyphenylpyruvate dioxygenase-like protein|Gloxd1|Glyoxalase domain containing 1|Glyoxalase domain-containing protein 1|Hpdl|HPDL_HUMAN
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:HPDL_HUMAN
- Purification:Antigen affinity purified
- Packaging:Vial
Spécifications
À propos de cet article
Type:
Antigen: HPDL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human