Anti-MED31 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MED31
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-970
- 77859-633
- Antigen symbol:
- MED31
- MED31
- Antigen synonyms:
- SOH1|CGI-125|Mediator of RNA polymerase II transcription subunit 31|Mediator complex subunit 31|Mediator complex subunit SOH1|hSOH1
- SOH1|CGI-125|Mediator of RNA polymerase II transcription subunit 31|Mediator complex subunit 31|Mediator complex subunit SOH1|hSOH1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3C7
- Q9Y3C7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mediator of RNA polymerase II transcription subunit 31 protein (2-72AA)
- Recombinant Human Mediator of RNA polymerase II transcription subunit 31 protein (2-72AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together