Anti-NAA20 Rabbit Polyclonal Antibody
Catalogue # 77743-959
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:NAA20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:P61599
- Antigen synonyms:NAA20|NAT5|N-alpha-acetyltransferase 20|EC 2.3.1.254|Methionine N-acetyltransferase|N-acetyltransferase 5|N-terminal acetyltransferase B complex catalytic subunit NAA20|N-terminal acetyltransferase B complex catalytic subunit NAT5|NatB complex subunit NAT5|NatB catalytic subunit
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:NAT5
- Purification:Antigen affinity purified
- Packaging:Vial
Spécifications
À propos de cet article
Type:
Antigen: NAA20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat