Anti-EPHA1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: EPHA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77854-788
- 77735-985
- Antigen symbol:
- EPHA1
- EPHA1
- Antigen synonyms:
- EPH|EPH receptor A1|EPH tyrosine kinase 1|EPH tyrosine kinase|EPHA1_HUMAN|Ephrin receptor Eph A1|Ephrin type A receptor 1|Ephrin type-A receptor 1|EPHT 1|EPHT|EPHT1|Erythropoietin producing hepatoma amplified sequence|Erythropoietin-producing hepatoma receptor|Esk|hMGC163163|Oncogene EPH|Soluble EPHA1 variant 1|Soluble EPHA1 variant 2|Tyrosine protein kinase receptor EPH|Tyrosine-protein kinase receptor EPH
- EPH|EPH receptor A1|EPH tyrosine kinase 1|EPH tyrosine kinase|EPHA1_HUMAN|Ephrin receptor Eph A1|Ephrin type A receptor 1|Ephrin type-A receptor 1|EPHT 1|EPHT|EPHT1|Erythropoietin producing hepatoma amplified sequence|Erythropoietin-producing hepatoma receptor|Esk|hMGC163163|Oncogene EPH|Soluble EPHA1 variant 1|Soluble EPHA1 variant 2|Tyrosine protein kinase receptor EPH|Tyrosine-protein kinase receptor EPH
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P21709
- P21709
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ephrin type-A receptor 1 protein (296-414AA)
- Recombinant Human Ephrin type-A receptor 1 protein (296-414AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together