Anti-KCNQ3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: KCNQ3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77741-527
- 77858-467
- Antigen symbol:
- KCNQ3
- KCNQ3
- Antigen synonyms:
- BFNC 2|BFNC|BFNC2|EBN 2|EBN2|KCNQ 3|KCNQ3_HUMAN|KQT like 3|KQT-like 3|KV7.3|Potassium channel subunit alpha KvLQT3|Potassium channel voltage gated subfamily Q member 3|Potassium voltage gated channel KQT like protein 3|Potassium voltage gated channel KQT like subfamily member 3|Potassium voltage gated channel subfamily KQT member 3|Potassium voltage-gated channel subfamily KQT member 3|Voltage gated potassium channel subunit Kv7.3|Voltage-gated potassium channel subunit Kv7.3
- BFNC 2|BFNC|BFNC2|EBN 2|EBN2|KCNQ 3|KCNQ3_HUMAN|KQT like 3|KQT-like 3|KV7.3|Potassium channel subunit alpha KvLQT3|Potassium channel voltage gated subfamily Q member 3|Potassium voltage gated channel KQT like protein 3|Potassium voltage gated channel KQT like subfamily member 3|Potassium voltage gated channel subfamily KQT member 3|Potassium voltage-gated channel subfamily KQT member 3|Voltage gated potassium channel subunit Kv7.3|Voltage-gated potassium channel subunit Kv7.3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43525
- O43525
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Potassium voltage-gated channel subfamily KQT member 3 protein (763-872AA)
- Recombinant Human Potassium voltage-gated channel subfamily KQT member 3 protein (763-872AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together