Anti-MRPL12 Rabbit Recombinant Polyclonal Antibody
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MRPL12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77743-390
- 77859-948
- Antigen symbol:
- MRPL12
- MRPL12
- Antigen synonyms:
- 39S ribosomal protein L12|39S ribosomal protein L12, mitochondrial|5c5-2|FLJ60124|L12mt|MGC8610|mitochondrial|Mitochondrial ribosomal protein L12|MRP-L12|MRP-L31/34|MRPL7|MRPL7/L12|RM12_HUMAN|RPML12
- 39S ribosomal protein L12|39S ribosomal protein L12, mitochondrial|5c5-2|FLJ60124|L12mt|MGC8610|mitochondrial|Mitochondrial ribosomal protein L12|MRP-L12|MRP-L31/34|MRPL7|MRPL7/L12|RM12_HUMAN|RPML12
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52815
- P52815
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human 39S ribosomal protein L12, mitochondrial protein (1-198AA)
- Recombinant Human 39S ribosomal protein L12, mitochondrial protein (1-198AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together