Anti-NTS Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:200 to 1:500
Type:
Antigen: NTS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77745-097
- 77745-098
- Antigen symbol:
- NTS
- NTS
- Antigen synonyms:
- Human proneurotensin|Large neuromedin N|Neuromedin N preproprotein|Neurotensin/neuromedin N|NEUT_HUMAN|NMN 125|NmN|NmN-125|NN|NT|NT/N|NTRH|NTS|NTS1|Pro neurotensin/neuromedin|Proneuromedin N mRNA|Tail peptide
- Human proneurotensin|Large neuromedin N|Neuromedin N preproprotein|Neurotensin/neuromedin N|NEUT_HUMAN|NMN 125|NmN|NmN-125|NN|NT|NT/N|NTRH|NTS|NTS1|Pro neurotensin/neuromedin|Proneuromedin N mRNA|Tail peptide
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P30990
- P30990
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Neurotensin/neuromedin N protein (24-143AA)
- Neurotensin/neuromedin N protein (24-143AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together