Anti-HMGN1 Rabbit Polyclonal Antibody
Catalogue # 77737-920
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:HMGN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:P05114
- Antigen synonyms:HMG14|Non-histone chromosomal protein HMG-14|High mobility group nucleosome-binding domain-containing protein 1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:High mobility group nucleosome-binding domain-containing protein 1
- Purification:Antigen affinity purified
- Packaging:Vial
Spécifications
À propos de cet article
Recommended dilution: IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA: 1:20000
Type:
Antigen: HMGN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat