Anti-LMNB2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LMNB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-045
- 77742-254
- Antigen symbol:
- LMNB2
- LMNB2
- Antigen synonyms:
- LAMB2|LMN 2|LMNB2_HUMAN|RGD1563803|LMN2|LMNB2|LAMB 2|MGC2721|LMN B2|Lamin-B2|LMNB 2
- LAMB 2|LAMB2|Lamin-B2|LMN 2|LMN B2|LMN2|LMNB 2|LMNB2|LMNB2_HUMAN|MGC2721|RGD1563803
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q03252
- Q03252
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lamin-B2 protein (442-615AA)
- Recombinant Human Lamin-B2 protein (442-615AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together