Anti-NMT2 Rabbit Recombinant Polyclonal Antibody
Catalogue # 77744-579
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:NMT2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:O60551
- Antigen synonyms:EC 2.3.1.97|Glycylpeptide N tetradecanoyltransferase 2|Glycylpeptide N-tetradecanoyltransferase 2|Myristoyl CoA:protein N myristoyltransferase 2|Myristoyl-CoA:protein N-myristoyltransferase 2|N-myristoyltransferase 2|NMT 2|NMT2_HUMAN|Peptide N myristoyltransferase 2|Peptide N-myristoyltransferase 2|Type II N myristoyltransferase|Type II N-myristoyltransferase
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Glycylpeptide N-tetradecanoyltransferase 2 protein (1-120AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Spécifications
À propos de cet article
Recommended dilution: IHC: 1:20 to 1:200, IP:1:200 to 1:2000
Type:
Antigen: NMT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human