Anti-F11 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: F11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77855-028
- 77736-333
- Antigen symbol:
- F11
- F11
- Antigen synonyms:
- coagulation factor XI|Coagulation factor XIa light chain|FA11_HUMAN|FXI|MGC141891|Plasma thromboplastin antecedent|Platelet coagulation factor XI|PTA
- coagulation factor XI|Coagulation factor XIa light chain|FA11_HUMAN|FXI|MGC141891|Plasma thromboplastin antecedent|Platelet coagulation factor XI|PTA
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P03951
- P03951
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Coagulation factor XI protein (19-387AA)
- Recombinant Human Coagulation factor XI protein (19-387AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together