Anti-GNB5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GNB5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-581
- 77856-210
- Antigen symbol:
- GNB5
- GNB5
- Antigen synonyms:
- FLJ37457|FLJ43714|G protein beta 5 subunit|G protein beta subunit 5L|GB 5|GB5|GBB5_HUMAN|Gbeta5|GNB 5|GNB5|Guanine nucleotide binding protein (G protein) beta 5|Guanine nucleotide binding protein beta 5|Guanine nucleotide binding protein beta 5 subunit|Guanine nucleotide binding protein beta subunit 5L|Guanine nucleotide binding protein subunit beta 5|Guanine nucleotide-binding protein subunit beta-5|Transducin beta chain 5
- FLJ43714|Guanine nucleotide binding protein beta 5 subunit|Guanine nucleotide binding protein subunit beta 5|G protein beta 5 subunit|Guanine nucleotide binding protein beta 5|FLJ37457|GB 5|GB5|Guanine nucleotide binding protein (G protein) beta 5|Transducin beta chain 5|GBB5_HUMAN|GNB 5|Guanine nucleotide binding protein beta subunit 5L|G protein beta subunit 5L|Gbeta5|Guanine nucleotide-binding protein subunit beta-5|GNB5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14775
- O14775
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Guanine nucleotide-binding protein subunit beta-5 protein (1-353AA)
- Recombinant Human Guanine nucleotide-binding protein subunit beta-5 protein (1-353AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together