Order Entry
Canada
ContactUsLinkComponent
Anti-MTSS1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur:  AFG Bioscience
New Product
Certificats
undefined
Anti-MTSS1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur:  AFG Bioscience
New Product

Spécifications

  • Catalog No:
  • 77860-115
  • 77743-626
  • Antigen symbol:
  • MTSS1
  • MTSS1
  • Antigen synonyms:
  • 2310003N14Rik|BC024131|D130001D01Rik|DKFZp781P2223|FLJ44694|KIAA0429|metastasis suppressor 1|Metastasis suppressor protein 1|Metastasis suppressor YGL 1|Metastasis suppressor YGL-1|MGC37896|MIM|MIMA|MIMB|missing in metastasis|Missing in metastasis protein|mKIAA0429|MTSS 1|MTSS1_HUMAN
  • 2310003N14Rik|mKIAA0429|DKFZp781P2223|FLJ44694|Metastasis suppressor YGL 1|Missing in metastasis protein|missing in metastasis|BC024131|MIM|D130001D01Rik|Metastasis suppressor protein 1|MTSS 1|MIMA|MIMB|metastasis suppressor 1|Metastasis suppressor YGL-1|MTSS1_HUMAN|MGC37896|KIAA0429
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O43312
  • O43312
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Metastasis suppressor protein 1 protein (616-730AA)
  • Recombinant Human Metastasis suppressor protein 1 protein (616-730AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Spécifications

Frequently Bought Together