Anti-OPHN1 Rabbit Recombinant Polyclonal Antibody
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: OPHN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77745-507
- 77745-506
- Antigen symbol:
- OPHN1
- OPHN1
- Antigen synonyms:
- OPHN1|Oligophrenin-1
- OPHN1|Oligophrenin-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60890
- O60890
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Oligophrenin-1 protein (1-270AA)
- Oligophrenin-1 protein (1-270AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together