Anti-MOGAT2 Rabbit Polyclonal Antibody
Catalogue # 77743-301
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:MOGAT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q3SYC2
- Antigen synonyms:2 acylglycerol O acyltransferase 2|2-acylglycerol O-acyltransferase 2|Acyl CoA:monoacylglycerol acyltransferase 2|Acyl-CoA:monoacylglycerol acyltransferase 2|DC5|DGAT2L5|Diacylglycerol acyltransferase 2 like protein 5|Diacylglycerol acyltransferase 2-like protein 5|Diacylglycerol O acyltransferase candidate 5|Diacylglycerol O-acyltransferase candidate 5|EC 2.3.1.22|FLJ22644|hDC5|hMGAT2|Mgat1l|MGAT2|MGC119183|MGC119184|MGC119185|MGC189143|MOGT2_HUMAN|Monoacylglycerol O acyltransferase 1 like|Monoacylglycerol O acyltransferase 2|Monoacylglycerol O-acyltransferase 2
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Monoacylglycerol O-acyltransferase 2
- Purification:Antigen affinity purified
- Packaging:Vial
Spécifications
À propos de cet article
Type:
Antigen: MOGAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat