Anti-Lypla1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: Lypla1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
- Catalog No:
- 77742-506
- 77859-270
- Antigen symbol:
- Lypla1
- Lypla1
- Antigen synonyms:
- Lypla1|Apt1|Pla1a|Acyl-protein thioesterase 1|APT-1|EC 3.1.2.-|Lysophospholipase 1|Lysophospholipase I|LPL-I|LysoPLA I
- Pla1a|EC 3.1.2.-|Lysophospholipase 1|Lypla1|Acyl-protein thioesterase 1|Lysophospholipase I|LPL-I|LysoPLA I|APT-1|Apt1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P97823
- P97823
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Mouse Acyl-protein thioesterase 1 protein (1-230AA)
- Recombinant Mouse Acyl-protein thioesterase 1 protein (1-230AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together