Anti-RTN4RL2 Rabbit Polyclonal Antibody
50 µl, Vial
77832-679
: A59854-50UL
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: RTN4RL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Taille:50 µl
- Packaging:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- ImmunoFluorescence:Yes
- Isotype:IgG
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Host:Rabbit
- Shipping temperature:Shipped on Blue ice
- Antigen name:Reticulon-4 receptor-like 2 protein (355-372AA)
- Cat. No.:77832-679
- Reactivity:Human
- Gene ID:Q86UN3
- Antigen synonyms:RTN4RL2| NGRH1| NGRL3| Reticulon-4 receptor-like 2| Nogo receptor-like 3| Nogo-66 receptor homolog 1| Nogo-66 receptor-related protein 2| NgR2
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:RTN4RL2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



