Anti-C2CD6 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: C2CD6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-777
- 77826-778
- Antigen symbol:
- C2CD6
- C2CD6
- Antigen name:
- C2 calcium-dependent domain-containing protein 6 protein (401-623AA)
- C2 calcium-dependent domain-containing protein 6 protein (401-623AA)
- Antigen synonyms:
- C2CD6|ALS2CR11C2 calcium-dependent domain-containing protein 6|Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 11 protein
- C2CD6|ALS2CR11C2 calcium-dependent domain-containing protein 6|Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 11 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q53TS8
- Q53TS8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together