Anti-TRERF1 Rabbit Polyclonal Antibody
Catalog # 77803-509
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Zinc finger transcription factor TReP-132
- Antigen symbol:TRERF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96PN7
- Antigen synonyms:TRERF1|BCAR2|RAPA|TREP132|Transcriptional-regulating factor 1|Breast cancer anti-estrogen resistance 2|Transcriptional-regulating protein 132|Zinc finger protein rapa|Zinc finger transcription factor TReP-132
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TRERF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse