Anti-CYFIP1 Rabbit Recombinant Polyclonal Antibody (FITC)
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: CYFIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77830-205
- 77830-204
- Antigen symbol:
- CYFIP1
- CYFIP1
- Antigen name:
- Cytoplasmic FMR1-interacting protein 1 protein (320-449AA)
- Cytoplasmic FMR1-interacting protein 1 protein (320-449AA)
- Antigen synonyms:
- CY|CYFIP 1|CYFIP1|CYFP1_HUMAN|Cytoplasmic FMR1 interacting protein 1|Cytoplasmic FMR1-interacting protein 1|FLJ45151|KIAA0068|p140sra 1|p140sra-1|P140SRA1|Selective hybridizing clone|SHYC|Specifically Rac1 associated protein 1|Specifically Rac1-associated protein 1|SRA 1|Sra-1|SRA1
- CY|CYFIP 1|CYFIP1|CYFP1_HUMAN|Cytoplasmic FMR1 interacting protein 1|Cytoplasmic FMR1-interacting protein 1|FLJ45151|KIAA0068|p140sra 1|p140sra-1|P140SRA1|Selective hybridizing clone|SHYC|Specifically Rac1 associated protein 1|Specifically Rac1-associated protein 1|SRA 1|Sra-1|SRA1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7L576
- Q7L576
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together