Anti-TRNAU1AP Rabbit Recombinant Polyclonal Antibody
77841-054
New Product
- Antigen name:tRNA selenocysteine 1-associated protein 1 protein (33-156AA)
- Antigen symbol:TRNAU1AP
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NX07
- Antigen synonyms:1110007F05Rik|AA387999|FLJ20503|PRO1902|RP4 669K10.4|SECp43|tRNA selenocysteine 1-associated protein 1|tRNA selenocysteine associated protein 1|tRNA selenocysteine-associated protein 1|Trnau1ap|TRSPAP1|TSAP1_HUMAN|WI1 2643G14.2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: TRNAU1AP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human