Anti-TMPO Rabbit Polyclonal Antibody
Catalog # 77803-175
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:TP5)]
- Antigen symbol:TMPO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P42166
- Antigen synonyms:TMPO| LAP2Lamina-associated polypeptide 2| isoform alpha| Thymopoietin isoform alpha| TP alpha| Thymopoietin-related peptide isoform alpha| TPRP isoform alpha) [Cleaved into: Thymopoietin| TP| Splenin)| Thymopentin| TP5)]
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TMPO
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat