Anti-PBXIP1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PBXIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-930
- 77833-929
- Antigen symbol:
- PBXIP1
- PBXIP1
- Antigen name:
- Pre-B-cell leukemia transcription factor-interacting protein 1 protein (361-600AA)
- Pre-B-cell leukemia transcription factor-interacting protein 1 protein (361-600AA)
- Antigen synonyms:
- PBXIP1|HPIPPre-B-cell leukemia transcription factor-interacting protein 1|Hematopoietic PBX-interacting protein
- PBXIP1|HPIPPre-B-cell leukemia transcription factor-interacting protein 1|Hematopoietic PBX-interacting protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96AQ6
- Q96AQ6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together