Anti-Pcsk1 Rabbit Recombinant Polyclonal Antibody
New Product
Type:
Antigen: Pcsk1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
- Catalog No:
- 77793-447
- 77793-446
- Antigen symbol:
- Pcsk1
- Pcsk1
- Antigen name:
- Recombinant Rat Neuroendocrine convertase 1 protein (111-752AA)
- Recombinant Rat Neuroendocrine convertase 1 protein (111-752AA)
- Antigen synonyms:
- Pcsk1|Bdp|Nec-1|Nec1Neuroendocrine convertase 1|NEC 1|EC 3.4.21.93|Prohormone convertase 1|Proprotein convertase 1|PC1
- Pcsk1|Bdp|Nec-1|Nec1Neuroendocrine convertase 1|NEC 1|EC 3.4.21.93|Prohormone convertase 1|Proprotein convertase 1|PC1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Rat
- Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P28840
- P28840
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together