Anti-CLVS2 Rabbit Polyclonal Antibody
77797-378
New Product
- Antigen name:RLBP1L2
- Antigen symbol:CLVS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q5SYC1
- Antigen synonyms:bA160A10.4|C6orf212|C6orf213|Clavesin-2|clvs2|CLVS2_HUMAN|Retinaldehyde-binding protein 1-like 2|RLBP1L2
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: CLVS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat