Anti-WDPCP Rabbit Recombinant Polyclonal Antibody (HRP)
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: WDPCP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77819-479
- 77819-480
- Antigen symbol:
- WDPCP
- WDPCP
- Antigen name:
- WD repeat-containing and planar cell polarity effector protein fritz homolog protein (621-737AA)
- WD repeat-containing and planar cell polarity effector protein fritz homolog protein (621-737AA)
- Antigen synonyms:
- Bardet-Biedl syndrome 15 protein|Chromosome 2 open reading frame 86|DKFZp686C12204|Fritz|FRITZ_HUMAN|hFrtz|WD repeat containing protein C2orf86|WD repeat-containing and planar cell polarity effector protein|WD repeat-containing and planar cell polarity effector protein fritz homolog|WDPCP
- Bardet-Biedl syndrome 15 protein|Chromosome 2 open reading frame 86|DKFZp686C12204|Fritz|FRITZ_HUMAN|hFrtz|WD repeat containing protein C2orf86|WD repeat-containing and planar cell polarity effector protein|WD repeat-containing and planar cell polarity effector protein fritz homolog|WDPCP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95876
- O95876
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together