Anti-DDX24 Rabbit Polyclonal Antibody
77808-350
New Product
- Antigen name:Synthesized peptide derived from internal of DDX24.
- Antigen symbol:DDX24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:Q9GZR7
- Antigen synonyms:ATP dependent RNA helicase DDX24|ATP-dependent RNA helicase ddx24|DDX 24|ddx24|DDX24_HUMAN|DEAD (Asp Glu Ala Asp) box polypeptide 24|DEAD box protein 24|DEAD/H (Asp Glu Ala Asp) box polypeptide 24|S. cerevisiae CHL1 like helicase
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:50 to 1:100
Type:
Antigen: DDX24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human