Anti-PEG10 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: PEG10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77831-699
- 77831-700
- Antigen symbol:
- PEG10
- PEG10
- Antigen name:
- Retrotransposon-derived protein PEG10 protein (1-325AA)
- Retrotransposon-derived protein PEG10 protein (1-325AA)
- Antigen synonyms:
- AA407948|Edr|Embryonal carcinoma differentiation regulated|Embryonal carcinoma differentiation-regulated protein|HB 1|HB1|KIAA1051|Mammalian retrotransposon-derived protein 2|Mar2|Mart2|MEF3 like 1|MEF3-like protein 1|MEF3L|MEF3L1|MyEF 3|Myelin expression factor 3-like protein 1|Paternally expressed 10|Paternally expressed gene 10 ORF1|Paternally expressed gene 10 protein|Peg10|PEG10 protein|PEG10_HUMAN|Putative uncharacterized protein PEG10|Retrotransposon gag domain containing 3|Retrotransposon gag domain-containing protein 3|Retrotransposon-derived gag-like polyprotein|Retrotransposon-derived protein PEG10|RGAG3|Ty3/Gypsy-like protein
- AA407948|Edr|Embryonal carcinoma differentiation regulated|Embryonal carcinoma differentiation-regulated protein|HB 1|HB1|KIAA1051|Mammalian retrotransposon-derived protein 2|Mar2|Mart2|MEF3 like 1|MEF3-like protein 1|MEF3L|MEF3L1|MyEF 3|Myelin expression factor 3-like protein 1|Paternally expressed 10|Paternally expressed gene 10 ORF1|Paternally expressed gene 10 protein|Peg10|PEG10 protein|PEG10_HUMAN|Putative uncharacterized protein PEG10|Retrotransposon gag domain containing 3|Retrotransposon gag domain-containing protein 3|Retrotransposon-derived gag-like polyprotein|Retrotransposon-derived protein PEG10|RGAG3|Ty3/Gypsy-like protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86TG7
- Q86TG7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together