Anti-FBXO44 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FBXO44
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-387
- 77838-386
- Antigen symbol:
- FBXO44
- FBXO44
- Antigen name:
- F-box only protein 44 protein (1-224AA)
- F-box only protein 44 protein (1-224AA)
- Antigen synonyms:
- 5730411K09|AV001623|DKFZp781J0852|F box gene 3|F box only protein 44|F box only protein 6a|F box protein 44|F box protein FBX30|F box/G domain protein 3|F-box only protein 44|F-box protein FBX30|F-box/G-domain protein 3|FBG3|FBX30|FBX44|FBX44_HUMAN|FBX6A|Fbxo44|Fbxo6a|MGC14140|RGD1562463
- 5730411K09|AV001623|DKFZp781J0852|F box gene 3|F box only protein 44|F box only protein 6a|F box protein 44|F box protein FBX30|F box/G domain protein 3|F-box only protein 44|F-box protein FBX30|F-box/G-domain protein 3|FBG3|FBX30|FBX44|FBX44_HUMAN|FBX6A|Fbxo44|Fbxo6a|MGC14140|RGD1562463
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H4M3
- Q9H4M3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together