Anti-SCOC Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: SCOC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77798-861
- 77798-862
- Antigen symbol:
- SCOC
- SCOC
- Antigen name:
- Short coiled-coil protein (1-121AA)
- Short coiled-coil protein (1-121AA)
- Antigen synonyms:
- HRIHFB2072| Scoc| SCOC_HUMAN| SCOCO| Short coiled coil protein| Short coiled-coil protein| UNC-69
- HRIHFB2072| Scoc| SCOC_HUMAN| SCOCO| Short coiled coil protein| Short coiled-coil protein| UNC-69
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UIL1
- Q9UIL1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together