Anti-PAK3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77793-162
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Serine/threonine-protein kinase PAK 3 protein (70-190AA)
- Antigen symbol:PAK3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O75914
- Antigen synonyms:PAK3|OPHN3|Serine/threonine-protein kinase PAK 3|EC 2.7.11.1|Beta-PAK|Oligophrenin-3|p21-activated kinase 3|PAK-3
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: PAK3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse