Anti-LRRN1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: LRRN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77830-129
- 77830-128
- Antigen symbol:
- LRRN1
- LRRN1
- Antigen name:
- Leucine-rich repeat neuronal protein 1 protein (26-631AA)
- Leucine-rich repeat neuronal protein 1 protein (26-631AA)
- Antigen synonyms:
- FIGLER3|KIAA1497|Leucine-rich repeat neuronal protein 1|LRRN1_HUMAN|Nbla10449|Neuronal leucine-rich repeat protein 1|NLRR-1
- FIGLER3|KIAA1497|Leucine-rich repeat neuronal protein 1|LRRN1_HUMAN|Nbla10449|Neuronal leucine-rich repeat protein 1|NLRR-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6UXK5
- Q6UXK5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together