Anti-TNFRSF10C Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: TNFRSF10C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77812-722
- 77812-721
- Antigen symbol:
- TNFRSF10C
- TNFRSF10C
- Antigen name:
- Tumor necrosis factor receptor superfamily member 10C protein (26-236AA)
- Tumor necrosis factor receptor superfamily member 10C protein (26-236AA)
- Antigen synonyms:
- DCR1|LIT|TRAILR3|TRID|UNQ321/PRO366|Tumor necrosis factor receptor superfamily member 10C|Antagonist decoy receptor for TRAIL/Apo-2L|Decoy TRAIL receptor without death domain|Decoy receptor 1|DcR1|Lymphocyte inhibitor of TRAIL|TNF-related apoptosis-inducing ligand receptor 3|TRAIL receptor 3|TRAIL-R3|TRAIL receptor without an intracellular domain|CD antigen CD263
- DCR1|LIT|TRAILR3|TRID|UNQ321/PRO366|Tumor necrosis factor receptor superfamily member 10C|Antagonist decoy receptor for TRAIL/Apo-2L|Decoy TRAIL receptor without death domain|Decoy receptor 1|DcR1|Lymphocyte inhibitor of TRAIL|TNF-related apoptosis-inducing ligand receptor 3|TRAIL receptor 3|TRAIL-R3|TRAIL receptor without an intracellular domain|CD antigen CD263
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14798
- O14798
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Spécifications
Frequently Bought Together