Anti-MAB21L1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: MAB21L1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77825-207
- 77825-206
- Antigen symbol:
- MAB21L1
- MAB21L1
- Antigen name:
- Putative nucleotidyltransferase MAB21L1 protein (1-359AA)
- Putative nucleotidyltransferase MAB21L1 protein (1-359AA)
- Antigen synonyms:
- MAB21L1|CAGR1|Nbla00126Putative nucleotidyltransferase MAB21L1|EC 2.7.7.-|Protein mab-21-like 1
- MAB21L1|CAGR1|Nbla00126Putative nucleotidyltransferase MAB21L1|EC 2.7.7.-|Protein mab-21-like 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13394
- Q13394
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together