Anti-CSRNP3 Rabbit Polyclonal Antibody
77809-284
New Product
- Antigen name:Synthesized peptide derived from the Internal region of CSRNP3.
- Antigen symbol:CSRNP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Monkey
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q8WYN3
- Antigen synonyms:CSRNP3|FAM130A2|TAIP2Cysteine/serine-rich nuclear protein 3|CSRNP-3|Protein FAM130A2|TGF-beta-induced apoptosis protein 2|TAIP-2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB:1:500 to 1:2000, ELISA:1:20000
Type:
Antigen: CSRNP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Monkey