Anti-CYFIP2 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA:1:2000 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: CYFIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
- Catalog No:
- 77810-775
- 77810-776
- Antigen symbol:
- CYFIP2
- CYFIP2
- Antigen name:
- Fusion protein of CYFIP2
- Fusion protein of CYFIP2
- Antigen synonyms:
- CYFIP2|KIAA1168|PIR121Cytoplasmic FMR1-interacting protein 2|p53-inducible protein 121
- CYFIP2|KIAA1168|PIR121Cytoplasmic FMR1-interacting protein 2|p53-inducible protein 121
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96F07
- Q96F07
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together