Anti-ZNF703 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZNF703
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77805-816
- 77805-817
- Antigen symbol:
- ZNF703
- ZNF703
- Antigen name:
- Zinc finger protein 703 protein (218-286AA)
- Zinc finger protein 703 protein (218-286AA)
- Antigen synonyms:
- FLJ14299| ZEPPO1| Zinc finger elbow-related proline domain protein 1| Zinc finger protein 703| ZN703_HUMAN| ZNF503L| znf703| ZPO1
- FLJ14299| ZEPPO1| Zinc finger elbow-related proline domain protein 1| Zinc finger protein 703| ZN703_HUMAN| ZNF503L| znf703| ZPO1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H7S9
- Q9H7S9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together