Anti-SYNCRIP Rabbit Polyclonal Antibody
77801-705
New Product
- Antigen name:cytoplasmic RNA-interacting protein
- Antigen symbol:SYNCRIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O60506
- Antigen synonyms:cytoplasmic RNA-interacting protein|dJ3J17.2|Glycine and tyrosine rich RNA binding protein|Glycine- and tyrosine-rich RNA-binding protein|GRY RBP|GRY-RBP|GRYRBP|Heterogeneous nuclear ribonucleoprotein Q|hnRNP Q|HNRPQ|HNRPQ_HUMAN|HNRPQ1|NS1 associated protein 1|NS1-associated protein 1|NSAP1|pp68|RP1 3J17.2|Synaptotagmin binding cytoplasmic RNA interacting protein|Synaptotagmin-binding|Syncrip
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: SYNCRIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat