Anti-MAP2K1/MAP2K2 Rabbit Polyclonal Antibody
Catalogue # 77806-420
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen name:Synthesized peptide derived from MEK-1/2 around the phosphorylation site of S222/226.
- Antigen symbol:MAP2K1/MAP2K2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q02750/P36507
- Antigen synonyms:MAP2K1|MEK1|PRKMK1|Dual specificity mitogen-activated protein kinase kinase 1|MAP kinase kinase 1|MAPKK 1|MKK1|ERK activator kinase 1|MAPK/ERK kinase 1|MEK 1|MAP2K2|MEK2|MKK2|PRKMK2|Dual specificity mitogen-activated protein k
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Spécifications
À propos de cet article
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:20000
Type:
Antigen: MAP2K1/MAP2K2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat