Anti-ATF6B Rabbit Polyclonal Antibody
77813-455
New Product
- Antigen name:Synthesized peptide derived from internal of ATF6B.
- Antigen symbol:ATF6B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q99941
- Antigen synonyms:ATF6B|CREBL1|G13Cyclic AMP-dependent transcription factor ATF-6 beta|cAMP-dependent transcription factor ATF-6 beta|Activating transcription factor 6 beta|ATF6-beta|Protein G13|cAMP response element-binding protein-related protein|Creb-rp|cAMP-responsive element-binding protein-like 1) [Cleaved into: Processed cyclic AMP-dependent transcription factor ATF-6 beta]
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: ATF6B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human