Anti-LYZ Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: LYZ
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77805-985
- 77805-984
- Antigen symbol:
- LYZ
- LYZ
- Antigen name:
- Lysozyme C protein (19-148AA)
- Lysozyme C protein (19-148AA)
- Antigen synonyms:
- 1 4 beta N acetylmuramidase C|1|4-beta-N-acetylmuramidase C|EC 3.2.1.17|LYSC_HUMAN|Lysosyme|Lysozyme (renal amyloidosis)|Lysozyme C|Lysozyme C precursor|LYZ|LZM|Renal amyloidosis
- 1 4 beta N acetylmuramidase C|1|4-beta-N-acetylmuramidase C|EC 3.2.1.17|LYSC_HUMAN|Lysosyme|Lysozyme (renal amyloidosis)|Lysozyme C|Lysozyme C precursor|LYZ|LZM|Renal amyloidosis
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P61626
- P61626
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together