Anti-CYP2U1 Rabbit Recombinant Polyclonal Antibody
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Recommended dilution: IF:1:50 to 1:200
Type:
Antigen: CYP2U1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77831-823
- 77831-824
- Antigen symbol:
- CP2U1_HUMAN;CYP2U 1;CYP2U1;Cytochrome P450 2U1;Cytochrome P450;family 2;subfamily U;polypeptide 1;EC 1.14.14.1;P450TEC
- CP2U1_HUMAN;CYP2U 1;CYP2U1;Cytochrome P450 2U1;Cytochrome P450;family 2;subfamily U;polypeptide 1;EC 1.14.14.1;P450TEC
- Antigen name:
- Cytochrome P450 2U1 protein (363-496AA)
- Cytochrome P450 2U1 protein (363-496AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z449
- Q7Z449
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together