Anti-MYOZ2 Rabbit Polyclonal Antibody
Catalogue # 77837-004
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen name:Synthetic peptide from protein at AA range: 21-70
- Antigen symbol:MYOZ2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NPC6
- Antigen synonyms:C4orf5|Calcineurin binding protein calsarcin 1|Calsarcin 1|Calsarcin-1|Calsarcin1|CMH16|CS 1|CS1|FATZ related protein 2|FATZ-related protein 2|Muscle specific protein|MYOZ 2|MYOZ2|MYOZ2_HUMAN|Myozenin-2|Myozenin2
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Spécifications
À propos de cet article
Recommended dilution: WB: 1:500 to 10000, ELISA: 1:10000
Type:
Antigen: MYOZ2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat