Anti-PCIF1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: PCIF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77842-053
- 77842-052
- Antigen symbol:
- PCIF1
- PCIF1
- Antigen name:
- Phosphorylated CTD-interacting factor 1 protein (201-500AA)
- Phosphorylated CTD-interacting factor 1 protein (201-500AA)
- Antigen synonyms:
- bA465L10.1| C20orf67| Pcif1| _HUMAN| PDX1 C terminal inhibiting factor 1| Phosphorylated CTD interacting factor 1| Phosphorylated CTD-interacting factor 1
- bA465L10.1| C20orf67| Pcif1| _HUMAN| PDX1 C terminal inhibiting factor 1| Phosphorylated CTD interacting factor 1| Phosphorylated CTD-interacting factor 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H4Z3
- Q9H4Z3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together